Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ F1142 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113412
Description
F1142 Polyclonal Antibody for Western Blot, IHC (P), ELISA
FAM114A2 (Family With Sequence Similarity 114 Member A2) is a Protein Coding gene. Diseases associated with FAM114A2 include Postural Orthostatic Tachycardia Syndrome and Phencyclidine Abuse. Among its related pathways are Oncogenic MAPK signaling and HIV Life Cycle. Gene Ontology (GO) annotations related to this gene include purine nucleotide binding. An important paralog of this gene is FAM114A1. [GeneCards].
Specifications
F1142 | |
Polyclonal | |
Unconjugated | |
FAM114A2 | |
133K02; 1810073G14Rik; 9030624B09Rik; C5orf3; Fam114a2; family with sequence similarity 114 member A2; family with sequence similarity 114, member A2; protein FAM114A2; testis tissue sperm-binding protein Li 81P | |
Rabbit | |
Protein G | |
RUO | |
10827, 67726 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
5 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q8VE88, Q9NRY5 | |
FAM114A2 | |
Recombinant Human Protein FAM114A2 protein (1-505AA). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction