Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
F13A1 Mouse anti-Human, Clone: F13A1/1683, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB14780
Description
Mouse monoclonal antibody raised against partial recombinant human F13A1.
- High specificity
- High reactivity
Specifications
| F13A1 | |
| Monoclonal | |
| Unconjugated | |
| ELISA Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| P00488 | |
| F13A1 | |
| Recombinant protein corresponding to amino acids 46-181 of human F13A1. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| F13A1/1683 | |
| Mouse monoclonal antibody raised against partial recombinant human F13A1. | |
| In 10 mM PBS. | |
| F13A | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 2162 | |
| Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction