Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

F13A1 Rabbit anti-Human, Unconjugated, Clone: F13A1/3772R, Abnova™

Catalog No. 89339503 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-503 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-503 Supplier Abnova Corporation Supplier No. RAB00547
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

F13A1 Monoclonal antibody specifically detects F13A1 in Human samples. It is validated for IHC-P.

Specifications

Antigen F13A1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone F13A1/3772R
Conjugate Unconjugated
Description coagulation factor XIII, A1 polypeptide
Gene F13A1
Gene Alias F13A
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 46-181 of human F13A1
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 2162
Target Species Human
Content And Storage Store at -20 to -80°C
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.