Learn More
Invitrogen™ F1A alpha Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA588764
Description
Immunogen sequence: LHALHLRQKG NRNTHKDLLR FAQVFSQMIH LNETVKAPDI ECVLRCSVLE IEQSMNRVKN ISDADVHNAM DNYECNLYTF LYLVCISTKT QCSEEDQCKI NKQIYNLIHL DPRTREGFTL LHLAVNSNTP VDDFHTNDVC SFPNALVTKL LLDCGAEVNA VDNEGNSALH IIVQYNRPIS DFLTLHSIII SLVEAGAHTD MTNKQNKTPL DKSTTGVSEI LLKTQMKMSL KCLAARAVRA NDINYQDQIP RTLEEFVGFH; Positive Samples: A-549; Cellular Location: Cytoplasm, Nucleus.
Fas and tumor necrosis factor receptor 1 (TNFR1) are two prototype members in the death receptor family. A novel protein that associates with the intracellular domains of Fas and TNFR1 was recently identified and designated F1Aalpha and FEM1-b. F1Aalpha/FEM1-b is the homologue of C. elegans sex determining protein FEM-1. FEM-1/F1Aalpha is cleaved by CED-3 and caspase. FEM-1/F1Aalpha associates with CED-4 and its mammalian homologue Apaf-1. Overexpression of F1Aalpha induces apoptosis. F1Aalpha is therefore a novel member of the death receptor associated protein that mediates apoptosis. F1Aalpha is expressed in a variety of human and mouse tissues.
Specifications
| F1A alpha | |
| Polyclonal | |
| Unconjugated | |
| FEM1B | |
| DKFZp451E0710; F1aa; F1A-alpha; fem-1 homolog B; fem-1 homolog b (C. elegans); Fem1b; FEM1-beta; FEM-1-like death receptor binding protein; fem-1-like death receptor-binding protein alpha; Fem-1-like in apoptotic pathway protein alpha; feminization 1 homolog b; feminization 1 homolog b (C. elegans); FIAA; Kiaa0396; mKIAA0396; mt-Fem; Protein fem-1 homolog B | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10116 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.56 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9UK73 | |
| FEM1B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 368-627 of human FEM1B (NP_0561371). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.