Learn More
Invitrogen™ F1A alpha Polyclonal Antibody

Description
F1A alpha Polyclonal Antibody for Western Blot, ELISA
Specifications
Specifications
| Antigen | F1A alpha |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 2.59 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Gene | FEM1B |
| Gene Accession No. | Q9UK73, Q9Z2G0 |
| Gene Alias | DKFZp451E0710; F1aa; F1A-alpha; fem-1 homolog B; fem-1 homolog b (C. elegans); Fem1b; FEM1-beta; FEM-1-like death receptor binding protein; fem-1-like death receptor-binding protein alpha; Fem-1-like in apoptotic pathway protein alpha; feminization 1 homolog b; feminization 1 homolog b (C. elegans); FIAA; Kiaa0396; mKIAA0396; mt-Fem; Protein fem-1 homolog B |
| Gene Symbols | FEM1B |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.