Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
F2 Mouse anti-Human, Clone: 1E12A8, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | F2 |
---|---|
Clone | 1E12A8 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6931
![]() ![]() |
Proteintech Group Inc
665091IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Prothrombin or Coagulation factor II, is proteolytically cleaved to form thrombin in the first step of the coagulation cascade which ultimately results in the stemming of blood loss. F2 also plays a role in maintaining vascular integrity during development and postnatal life. Mutations in F2 leads to various forms of thrombosis and dysprothrombinemia.Specifications
F2 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Coagulation factor II, F2, Prothrombin, PT | |
F2 | |
IgG1 | |
Protein G | |
Antibody |
1E12A8 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P00734 | |
2147 | |
F2 Fusion Protein Ag18385 | |
Primary | |
-20°C | |
F2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title