Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
F4/80 Antibody (521204), mFluor Violet 450 SE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB5580MFV450
Description
F4/80 Monoclonal antibody specifically detects F4/80 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
| F4/80 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Adaptive Immunity, Cellular Markers, Immune System Diseases, Immunology, Innate Immunity | |
| 13733 | |
| Store at 4C in the dark. |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 521204 | |
| Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| ADGRE1, cell surface glycoprotein F4/80, DD7A5-, DD7A5-7, EGF-like module containing, mucin-like, hormone receptor-like sequence 1, EGF-like module receptor 1, EGF-like module-containing mucin-like hormone receptor-like 1, EGF-TM7, Emr, Emr1, EMR1 hormone receptor, F4/80, Gpf480, Ly7, Ly71, lymphocyte antigen 71, Mouse EMR1, Rat EMR1, TM7L, TM7LN3 | |
| NS0 mouse myeloma cell line transfected with mouse F4/80/EMR1, Gln28-Gly931, Accession # Q61549 | |
| RUO | |
| Primary | |
| Mouse | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction