Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
F4/80 Antibody (CI-A3-1), Alexa Fluor™ 350, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB600404AF350
Description
F4/80 Monoclonal antibody specifically detects F4/80 in Human, Mouse, C. elegans, E. coli samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)Specifications
F4/80 | |
Monoclonal | |
Alexa Fluor 350 | |
ADGRE1, cell surface glycoprotein F4/80, DD7A5-, DD7A5-7, EGF-like module containing, mucin-like, hormone receptor-like sequence 1, EGF-like module receptor 1, EGF-like module-containing mucin-like hormone receptor-like 1, EGF-TM7, Emr, Emr1, EMR1 hormone receptor, F4/80, Gpf480, Ly7, Ly71, lymphocyte antigen 71, Mouse EMR1, Rat EMR1, TM7L, TM7LN3 | |
This F4/80 Antibody (CI-A3-1) was developed against Thioglycollate stimulated peritoneal macrophages from C57BL/6 mice. | |
0.125 mL | |
Adaptive Immunity, Cellular Markers, Immune System Diseases, Immunology, Innate Immunity | |
13733 | |
Store at 4°C in the dark. | |
IgG2b |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
CI-A3-1 | |
50mM Sodium Borate | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, C. elegans, E. coli | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction