Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
F4/80 Antibody (CI-A3-1), DyLight 488, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB600404G
Description
F4/80 Monoclonal specifically detects F4/80 in Human, Mouse, C. elegans, E. coli samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
F4/80 | |
Monoclonal | |
DyLight 488 | |
ADGRE1, Adhesion G protein-coupled receptor E1, cell surface glycoprotein F4/80, DD7A5-7, EGF-like module containing, mucin-like, hormone receptor-like sequence 1, EGF-like module receptor 1, EGF-like module-containing mucin-like hormone receptor-like 1, EGF-TM7, EMR1 hormone receptor, F4/80, Gpf480, Ly71, lymphocyte antigen 71, Mouse EMR1, Rat EMR1, TM7LN3 | |
Rat | |
160 kDa | |
0.125 mL | |
Immune System Diseases, Immunology, Innate Immunity | |
2015 | |
Human, Mouse, C. elegans, E. coli | |
Purified |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
CI-A3-1 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
ADGRE1 | |
Thioglycollate stimulated peritoneal macrophages from C57BL/6 mice. | |
Protein G purified | |
RUO | |
Primary | |
This recognizes mouse F4/80, a 160kD glycoprotein expressed by murine macrophages. | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction