Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
F4/80 Antibody (CI:A3-1), Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP261613SS
Description
F4/80 Monoclonal specifically detects F4/80 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry.Specifications
F4/80 | |
Monoclonal | |
Unconjugated | |
PBS with 0.02% Proclin 300 | |
Adgre1 | |
Thioglycollate stimulated peritoneal macrophages of mouse origin. | |
0.025 mg | |
Primary | |
This antibody recognises the mouse F4/80 antigen, a 160kD glycoprotein, expressed by murine macrophages. | |
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. | |
IgG1 κ |
Flow Cytometry, Immunohistochemistry | |
CI:A3-1 | |
Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500 | |
ADGRE1, Adhesion G protein-coupled receptor E1, cell surface glycoprotein F4/80, DD7A5-7, EGF-like module containing, mucin-like, hormone receptor-like sequence 1, EGF-like module receptor 1, EGF-like module-containing mucin-like hormone receptor-like 1, EGF-TM7, EMR1 hormone receptor, F4/80, Gpf480, Ly71, lymphocyte antigen 71, Mouse EMR1, Rat EMR1, TM7LN3 | |
Rat | |
Protein A purified | |
Immune System Diseases, Immunology, Innate Immunity, Myeloid derived Suppressor Cell | |
2015 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction