Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
F4/80 Antibody (Cl:A3-1 (recombinant version)), Alexa Fluor™ 405, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP281029AF405
Description
F4/80 Monoclonal specifically detects F4/80 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry.Specifications
F4/80 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Adgre1 | |
Thioglycollate stimulated peritoneal macrophages of mouse origin. | |
0.1 mL | |
Adaptive Immunity, GPCR, Immune System Diseases, Immunology, Innate Immunity, Myeloid derived Suppressor Cell | |
13733 | |
Mouse | |
Purified |
Flow Cytometry, Immunohistochemistry | |
Cl:A3-1 (recombinant version) | |
Flow Cytometry, Immunohistochemistry | |
ADGRE1, Adhesion G protein-coupled receptor E1, cell surface glycoprotein F4/80, DD7A5-7, EGF-like module containing, mucin-like, hormone receptor-like sequence 1, EGF-like module receptor 1, EGF-like module-containing mucin-like hormone receptor-like 1, EGF-TM7, EMR1 hormone receptor, F4/80, Gpf480, Ly71, lymphocyte antigen 71, Mouse EMR1, Rat EMR1, TM7LN3 | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
This antibody recognises the mouse F4/80 antigen, a 160kD glycoprotein, expressed by murine macrophages. | |
Store at 4°C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction