Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

F4/80-Like Receptor Rat anti-Mouse, Brilliant Violet 650, Clone: 6F12, BD Optibuild™

Catalog No. BDB744338 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB744338 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB744338 Supplier BD Biosciences Supplier No. 744338
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 6F12 antibody reacts with a 7-transmembrane-domain protein, which is similar to the F4/80 macrophage antigen of the EGF-TM7 protein family and is encoded by the Emr4 gene. The FIRE protein is expressed on myeloid cells with a denditic cell (DC) developmental potential, including subsets of DC and macrophages in the spleen and lymph nodes, most resident peritoneal macrophages, many peripheral blood monocytes, and a subpopulation of bone-marrow myeloid-cell progenitors. The protein is not detected on peripheral T and B lymphocytes, and it is down-regulated on thioglycollate-elicited peritoneal macrophages and on dendritic cells activated by GM-CSF, IFN-, anti-CD40, and LPS. Using soluble biotinylated fusion protein, a FIRE ligand was detected on a mouse IgG+ B lymphoma cell line (A20), but not on myeloid, fibroblast, or T-cell lines, suggesting that the FIRE protein may be involved in immunoregulatory interactions between antigen-presenting cells and B lymphocytes.

The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 650nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV650 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 650 nm. Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. BV650 will have moderate spillover into the BD Horizon™ BV711 detector.
TRUSTED_SUSTAINABILITY

Specifications

Antigen F4/80-Like Receptor
Classification Monoclonal
Clone 6F12
Concentration 0.2mg/mL
Conjugate Brilliant Violet 650
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Fire, Emr4, EGF-like module receptor 4, D17Ertd479e, Egf-tm7
Host Species Rat
Immunogen CHO cells expressing recombinant FIRE fusion protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.