Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

F4/80-Like Receptor Rat anti-Mouse, BUV395, Clone: 6F12, BD Optibuild™

Catalog No. BDB744341 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB744341 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB744341 Supplier BD Biosciences Supplier No. 744341
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 6F12 antibody reacts with a 7-transmembrane-domain protein, which is similar to the F4/80 macrophage antigen of the EGF-TM7 protein family and is encoded by the Emr4 gene. The FIRE protein is expressed on myeloid cells with a denditic cell (DC) developmental potential, including subsets of DC and macrophages in the spleen and lymph nodes, most resident peritoneal macrophages, many peripheral blood monocytes, and a subpopulation of bone-marrow myeloid-cell progenitors. The protein is not detected on peripheral T and B lymphocytes, and it is down-regulated on thioglycollate-elicited peritoneal macrophages and on dendritic cells activated by GM-CSF, IFN-, anti-CD40, and LPS. Using soluble biotinylated fusion protein, a FIRE ligand was detected on a mouse IgG+ B lymphoma cell line (A20), but not on myeloid, fibroblast, or T-cell lines, suggesting that the FIRE protein may be involved in immunoregulatory interactions between antigen-presenting cells and B lymphocytes.

The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348 nm and an Em Max at 395 nm, BD Horizon BUV395 can be excited with a 355 nm laser and detected with a 379/28 filter. BUV395 is a UV-excitable dye that has been developed exclusively by BD Biosciences. With an excitation max of 348 nm and emission max of 395 nm, BUV395 can be excited by the 355nm laser and detected in a 379/28 filter. This dye is optimal for multicolor flow cytometry because it has little to no spillover into other detectors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen F4/80-Like Receptor
Classification Monoclonal
Clone 6F12
Concentration 0.2mg/mL
Conjugate Brilliant Ultraviolet 395
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Fire, Emr4, EGF-like module receptor 4, D17Ertd479e, Egf-tm7
Host Species Rat
Immunogen CHO cells expressing recombinant FIRE fusion protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.