Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
F4/80 Rat anti-Mouse, Alexa Fluor 647, Clone: T45-2342, BD Pharmingen™
Rat Monoclonal Antibody
Supplier: BD Biosciences 565853
Description
F4/80 Monoclonal antibody specifically detects F4/80 in Mouse samples. It is validated for Flow cytometry.
F4/80 Monoclonal antibody specifically detects F4/80 in Mouse samples. It is validated for Flow cytometry.
Specifications
F4/80 | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
0.1 mg | |
Cell Biology | |
Mouse | |
Purified |
Flow Cytometry | |
T45-2342 | |
Alexa Fluor 647 | |
Gpf480; F480; Emr1; Ly71; DD7A5-7; EGF-TM7; TM7LN3 | |
Mouse F4/80 Recombinant Protein | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor™ 647 under optimum conditions and unreacted Alexa Fluor™ 647 was removed. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction