Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
F4/80 Rat anti-Mouse, BUV395, Clone: T45-2342, BD Horizon™
Rat Monoclonal Antibody
Supplier: BD Biosciences 565614
Description
F4/80 Monoclonal antibody specifically detects F4/80 in Mouse samples. It is validated for Flow cytometry.
Specifications
F4/80 | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Rat | |
50 μg | |
Cell Biology | |
Mouse | |
Purified |
Flow Cytometry | |
T45-2342 | |
Brilliant Ultraviolet 395 | |
Gpf480; F480; Emr1; Ly71; DD7A5-7; EGF-TM7; TM7LN3 | |
Mouse F4/80 Recombinant Protein | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity and unconjugated antibody and free BD Horizon™ BUV395 were removed. chromatography. The antibody was conjugated with BD Horizon™ BUV395 under optimum conditions | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Reseach Use Only.
Spot an opportunity for improvement?Share a Content Correction