Learn More
Epredia™ Series 215 Laser Biopsy Cassettes with Lids
Catalog No. 2150510LM Shop All Epredia Products
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Pink
Red
White
Yellow
This item is not returnable. View return policy
This item is not returnable. View return policy
Manufactured specifically for laser printing, the Epredia™ Series 215 Biopsy Cassettes come with separate lids and are designed for small tissue samples.
  • Epredia™ Laser Cassettes are designed for use on both the NOVA™ and VEGA™ Laser Printers.
  • They contain a special constituent chemical compound that carbonizes when exposed to the laser beam.
  • Each style comes in 10 different colors: white, yellow, blue, pink, green, orange, grey, aqua, lilac and red with color-matched lids.
  • Each cassette style is designed to maximize fluid exchange during processing and come in stacks held together with tape for ease of loading into Epredia™ cassette hoppers for printing.
  • Series 110, 115 and 215 come in taped stacks of 40 cassettes; 25 stacks per case.
  • Series 815 come in taped stacks of 40 cassettes with lids attached; 45 stacks per case.
  • The Epredia™ Laser Cassette is not compatible with thermal tape style printers.
Specifications
Pore Style Small Squares
Quantity 40 Cassettes/Taped Stack, 25 Stacks/Cs.
Closure Included Yes
Closure Type Separate Lid
Color Green
Product Type Cassette
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.