Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FA156 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563337
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563337 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563337 Supplier Invitrogen™ Supplier No. PA563337
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSSWETLVQG LSGLTLSLGT NQPGPLPEAA LQPQETEEKR QRERQQESKI MFQRLLKQWL EE Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 63%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen FA156
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM156A
Gene Accession No. Q8NDB6
Gene Alias FAM156A; FAM156B; family with sequence similarity 156 member A; family with sequence similarity 156, member A; PP12994; PRO0659; protein FAM156A; protein FAM156A/FAM156B; TMEM29; TMEM29B; transmembrane protein 29; transmembrane protein 29/29B
Gene Symbols FAM156A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FA156. Recombinant protein control fragment (Product #RP-104511).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29057
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.