Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FA156 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113248
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113248 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113248 Supplier Thermo Scientific Supplier No. PA5113248
Add to Cart
Add to Cart

FA156 Polyclonal Antibody

FA156 Polyclonal Antibody for Western Blot, IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen FA156
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene FAM156A
Gene Accession No. Q8NDB6
Gene Alias FAM156A; FAM156B; family with sequence similarity 156 member A; family with sequence similarity 156, member A; PP12994; PRO0659; protein FAM156A; protein FAM156A/FAM156B; TMEM29; TMEM29B; transmembrane protein 29; transmembrane protein 29/29B
Gene Symbols FAM156A
Host Species Rabbit
Immunogen Recombinant Human Protein FAM156A/FAM156B protein (1-153AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29057
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.