Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FA2H Antibody (OTI2E3), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272192F
Description
FA2H Monoclonal specifically detects FA2H in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
FA2H | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
FA2H | |
Full length recombinant protein of human FA2H (NP_077282) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Immunocytochemistry, Immunofluorescence | |
OTI2E3 | |
Immunocytochemistry/Immunofluorescence | |
FAAHFAXDC1, FAH1, fatty acid 2-hydroxylase, Fatty acid alpha-hydroxylase, fatty acid hydroxylase domain containing 1, FLJ25287, SCS7, spastic paraplegia 35 (autosomal recessive), SPG35 | |
Mouse | |
Affinity Purified | |
Lipid and Metabolism | |
79152.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction