Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FA2H Monoclonal Antibody (OTI1B5), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA506832

Description
FA2H is a protein that catalyzes the synthesis of 2-hydroxysphingolipids, a subset of sphingolipids that contain 2-hydroxy fatty acids. Sphingolipids play roles in many cellular processes and their structural diversity arises from modification of the hydrophobic ceramide moiety, such as by 2-hydroxylation of the N-acyl chain, and the existence of many different head groups.Specifications
FA2H | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q7L5A8 | |
FA2H | |
Full length human recombit protein of human FA2H produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI1B5 | |
Unconjugated | |
FA2H | |
FAAH, FAH1, FAXDC1, SCS7, SPG35 | |
Mouse | |
Affinity Chromatography | |
RUO | |
79152 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction