Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FA2H Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100512
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100512 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100512 Supplier Invitrogen™ Supplier No. PA5100512
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total FA2H.

FA2H is a protein that catalyzes the synthesis of 2-hydroxysphingolipids, a subset of sphingolipids that contain 2-hydroxy fatty acids. Sphingolipids play roles in many cellular processes and their structural diversity arises from modification of the hydrophobic ceramide moiety, such as by 2-hydroxylation of the N-acyl chain, and the existence of many different head groups.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FA2H
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FA2H
Gene Accession No. Q2LAM0, Q5MPP0, Q7L5A8
Gene Alias FA2H; FAAH; FAH1; Fatty acid 2-hydroxylase; fatty acid alpha-hydroxylase; fatty acid hydroxylase domain containing 1; Faxdc1; FLJ25287; G630055L08Rik; RGD1310347; SCS7; spastic paraplegia 35 (autosomal recessive); SPG35; WD repeat domain 59; Wdr59
Gene Symbols FA2H
Host Species Rabbit
Immunogen A synthesized peptide derived from human FA2H(Accession Q7L5A8), corresponding to amino acid residues L33-G83.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 307855, 338521, 79152
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.