Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM53C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560125
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560125 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560125 Supplier Invitrogen™ Supplier No. PA560125
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WRPRGLRNLP RSRSQPCDLD ARKTGVKRRH EEDPRRLRPS LDFDKMNQKP YSGGLCLQET AREGSSISPP WFMACSPPPL SASCSPTGGS SQVLSESEE Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 96%.

The protein encoded by this gene belongs to the FAM53 protein family. FAM53 protein family members bind to a transcriptional regulator that modulates cell proliferation. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM53C
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM53C
Gene Accession No. Q9NYF3
Gene Alias 2810012G03Rik; C5orf6; FAM53C; family with sequence similarity 53 member C; family with sequence similarity 53, member C; Gm21179; Protein FAM53C; putative nuclear protein
Gene Symbols FAM53C
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FA53C. Recombinant protein control fragment (Product #RP-100131).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51307
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.