Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAAH Recombinant Rabbit Monoclonal Antibody (8G3)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA549927
Description
FAAH Recombinant Monoclonal Antibody for Western Blot, IHC (P), ELISA
Degrades bioactive fatty acid amides like oleamide, the endogenous levels of cannabinoid, anandamide and myristic amide to their corresponding acids, thereby serving to terminate the signaling functions of these molecules. Hydrolyzes polyunsaturated substrate anandamide preferentially as compared to monounsaturated substrates.
Specifications
FAAH | |
Recombinant Monoclonal | |
2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O00519, O08914 | |
FAAH | |
A synthesized peptide derived from human FAAH1. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
8G3 | |
Unconjugated | |
FAAH | |
amide hydrolase; anandamide amidohydrolase 1; AW412498; FA2H; Faah; FAAH1; FAAH-1; fatty acid amide hydrolase; fatty acid amide hydrolase protein; fatty-acid amide hydrolase 1; fatty-acid amide hydrolase 1-like; LOC100911581; MGC102823; MGC138146; Oleamide hydrolase; Oleamide hydrolase 1; PSAB | |
Rabbit | |
Affinity chromatography | |
RUO | |
14073, 2166 | |
-20°C or -80°C if preferred | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction