Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAAH2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106978
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106978 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106978 Supplier Invitrogen™ Supplier No. PA5106978
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAAH2 Polyclonal Antibody for Western Blot

FAAH2 encodes a fatty acid amide hydrolase that shares a conserved protein motif with the amidase signature family of enzymes. The encoded enzyme is able to catalyze the hydrolysis of a broad range of bioactive lipids, including those from the three main classes of fatty acid amides; N-acylethanolamines, fatty acid primary amides and N-acyl amino acids. This enzyme has a preference for monounsaturated acyl chains as a substrate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAAH2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene FAAH2
Gene Accession No. Q6GMR7
Gene Alias AMDD; amidase domain containing; amidase domain-containing protein; Anandamide amidohydrolase 2; FAAH2; fatty acid amide hydrolase 2; fatty-acid amide hydrolase 2; Oleamide hydrolase 2
Gene Symbols FAAH2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal region of human FAAH2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 158584
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.