Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAAP100 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP215635
Description
FAAP100 Polyclonal specifically detects FAAP100 in Human samples. It is validated for Western Blot.Specifications
FAAP100 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0) and 20% Glycerol with 0.01% Thimerosal | |
C17ORF70 | |
Recombinant protein encompassing a sequence within the center region of human FAAP100. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
80233 | |
Human | |
IgG |
Western Blot | |
1mg/mL | |
Western Blot 1:500-1:3000 | |
C17orf70, chromosome 17 open reading frame 70, FAAP100, Fanconi anemia-associated protein of 100 kDa, FLJ22175, FLJ30151 | |
Rabbit | |
93 kDa | |
0.1 mL | |
Primary | |
Human. Expected to cross react based on sequence identity: Monkey (98%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction