Learn More
Invitrogen™ FAAP24 Polyclonal Antibody

Description
Immunogen sequence: VLDLGMVLLP VASQMEASCL VIQLVQEQTK EPSKNPLLGK KRALLLSEPS LLRTVQQIPG VGKVKAPLLL QKFPSIQQLS NASIGELEQV VGQAVAQQ Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.
Specifications
Specifications
| Antigen | FAAP24 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | FAAP24 |
| Gene Accession No. | Q9BTP7 |
| Gene Alias | C19orf40; FAAP24; Fanconi anemia core complex associated protein 24; Fanconi anemia core complex-associated protein 24; Fanconi anemia-associated protein of 24 kDa |
| Gene Symbols | FAAP24 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.