Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAAP24 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA530903
Description
Recommended positive controls: 293T, A431, Jurkat, Raji. Predicted reactivity: Mouse (80%), Rat (80%), Bovine (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
FAAP24 is a component of the Fanconi anemia (FA) core complex (see MIM 227650), which plays a crucial role in DNA damage response (Ciccia et al., 2007 [PubMed 17289582]).
Specifications
FAAP24 | |
Polyclonal | |
Unconjugated | |
FAAP24 | |
C19orf40; FAAP24; Fanconi anemia core complex associated protein 24; Fanconi anemia core complex-associated protein 24; Fanconi anemia-associated protein of 24 kDa | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
91442 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.94 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9BTP7 | |
FAAP24 | |
Recombinant fragment corresponding to a region within amino acids 20 and 215 of Human FAAP24. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction