Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAAP24 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | FAAP24 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
FAAP24 Polyclonal specifically detects FAAP24 in Human samples. It is validated for Western Blot.Specifications
FAAP24 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
91442 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
chromosome 19 open reading frame 40, FAAP24FLJ46828, Fanconi anemia-associated protein of 24 kDa, Fanconi anemia-associated protein, 24 kDa, MGC32020 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-215 of human FAAP24 (NP_689479.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title