Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FABP1/L-FABP Antibody (FABP1/3482), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308934AF647
Description
FABP1/L-FABP Monoclonal antibody specifically detects FABP1/L-FABP in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin), Protein ArraySpecifications
FABP1/L-FABP | |
Monoclonal | |
Alexa Fluor 647 | |
FABPL, fatty acid binding protein 1, liver, fatty acid-binding protein, liver, L-FABPFatty acid-binding protein 1, Liver-type fatty acid-binding protein | |
Human recombinant FABP1/L-FABP protein fragment (around aa1-127) (exact sequence is proprietary) (Uniprot: P07148) | |
100 μg | |
Breast Cancer, Cancer, Cholesterol Metabolism, Diabetes Research, Lipid and Metabolism | |
2168 | |
Store at 4°C in the dark. | |
IgG2b κ |
Western Blot, Immunohistochemistry (Paraffin), Protein Array | |
FABP1/3482 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction