Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FABP1/L-FABP Rabbit anti-Human, Mouse, Rat, Clone: 8Q8S10, Novus Biologicals™

Rabbit Monoclonal Antibody
$206.50 - $492.50
Specifications
Antigen | FABP1/L-FABP |
---|---|
Clone | 8Q8S10 |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
FABP1/L-FABP Monoclonal antibody specifically detects FABP1/L-FABP in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
FABP1/L-FABP | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Breast Cancer, Cancer, Cholesterol Metabolism, Diabetes Research, Lipid and Metabolism | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
2168 | |
IgG | |
Affinity purified |
8Q8S10 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
FABPL, fatty acid binding protein 1, liver, fatty acid-binding protein, liver, L-FABPFatty acid-binding protein 1, Liver-type fatty acid-binding protein | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human FABP1/L-FABP (P07148). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title