Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FABP1 Recombinant Rabbit Monoclonal Antibody (8Q8S10)
GREENER_CHOICE

Catalog No. PIMA535080
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535080 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535080 Supplier Invitrogen™ Supplier No. MA535080
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

FABP1 Recombinant Monoclonal Antibody for ELISA, ICC/IF, Western Blot

FABP1 encodes the fatty acid binding protein found in liver. Fatty acid binding proteins are a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. FABP1 and FABP6 (the ileal fatty acid binding protein) are also able to bind bile acids. It is thought that FABPs roles include fatty acid uptake, transport, and metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FABP1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 8Q8S10
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide; pH 7.3
Gene FABP1
Gene Accession No. P02692, P07148, P12710
Gene Alias 14 kDa selenium-binding protein; FABP1; FABPL; Fabplg; fatty acid binding protein 1; fatty acid binding protein 1, liver; fatty acid binding protein liver; fatty acid-binding protein 1; Fatty acid-binding protein, liver; L-FABP; Liver-type fatty acid-binding protein; p14; SCP; SCP.; Squalene- and sterol-carrier protein; Z-protein
Gene Symbols FABP1
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14080, 2168, 24360
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.