Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FABP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547159
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547159 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547159 Supplier Invitrogen™ Supplier No. PA547159
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In Western blots, less than 5% cross-reactivity with recombinant rat FABP2, recombinant human FABP3, recombinant mouse (rm) FABP4, and rmFABP5 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

FABP1 encodes the fatty acid binding protein found in liver. Fatty acid binding proteins are a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. FABP1 and FABP6 (the ileal fatty acid binding protein) are also able to bind bile acids. It is thought that FABPs roles include fatty acid uptake, transport, and metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FABP1
Applications Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene FABP1
Gene Accession No. P02692, P07148, P12710
Gene Alias 14 kDa selenium-binding protein; FABP1; FABPL; Fabplg; fatty acid binding protein 1; fatty acid binding protein 1, liver; fatty acid binding protein liver; fatty acid-binding protein 1; Fatty acid-binding protein, liver; L-FABP; Liver-type fatty acid-binding protein; p14; SCP; SCP.; Squalene- and sterol-carrier protein; Z-protein
Gene Symbols FABP1
Host Species Goat
Immunogen E. coli-derived recombinant rat FABP1/L-FABP Met1-Ile127.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14080, 2168, 24360
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.