Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FABP10A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572183
Description
FABP10A Polyclonal Antibody for Western Blot
Binds hydrophobic ligands, such as cholate, in the cytoplasm. May be involved in intracellular lipid transport. Binds one cholate per subunit.
Specifications
FABP10A | |
Polyclonal | |
Unconjugated | |
fabp10a | |
fabp10; fabp10a; fatty acid binding protein 10, liver basic; fatty acid binding protein 10a, liver basic; fatty acid-binding protein 10; fatty acid-binding protein 10-A, liver basic; Fatty acid-binding protein, liver; L-BABP; Lb-FABP; Lfabp; L-FABP; liver bile acid-binding protein; liver-basic fatty acid binding protein; liver-type FABP; liver-type fatty acid binding protein; Liver-type fatty acid-binding protein; small cytosolic protein involved in lipid transport and metabolism; Zf-FABP10; zf-Lb-FABP; zgc:103719; zgc:92741; z-L-BABP | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
171481 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.46 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q9I8L5 | |
fabp10a | |
KLH conjugated synthetic peptide between 47-80 amino acids of DANRE fabp10a. | |
400 μL | |
Primary | |
Zebrafish | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction