Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FABP12 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576451
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
May play a role in lipid transport. [UniProt].
Specifications
FABP12 | |
Polyclonal | |
Unconjugated | |
FABP12 | |
1700008G05Rik; Fabp12; FABP12 {ECO:0000303; fatty acid binding protein 12; fatty acid binding protein ORF; fatty acid-binding protein 12; fatty acid-binding protein 12 {ECO:0000303; intracellular fatty acid-binding protein FABP12; Probable fatty acid-binding protein ENSP00000353650; probable fatty acid-binding protein ENSP00000353650 homolog; PubMed:18786628}; RGD1565000 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
499570, 646486 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
A6NFH5, B7SUM8 | |
FABP12 | |
Recombinant fusion protein of human FABP12. | |
100 μL | |
Primary | |
Human, Rat, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction