Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FABP2/I-FABP Antibody (9A9B7B3), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP151589
Description
FABP2/I-FABP Monoclonal specifically detects FABP2/I-FABP in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
FABP2/I-FABP | |
Monoclonal | |
Unconjugated | |
FABPI, fatty acid binding protein 2, intestinal, Fatty acid-binding protein 2, fatty acid-binding protein, intestinal, I-FABPMGC133132, Intestinal-type fatty acid-binding protein | |
Mouse | |
15 kDa | |
0.1 mL | |
Cholesterol Metabolism, Lipid and Metabolism | |
2169 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
9A9B7B3 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
FABP2 | |
Purified recombinant fragment of human FABP2 expressed in E. Coli. | |
Unpurified | |
RUO | |
Primary | |
Human, Mouse | |
Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction