Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FABP2/I-FABP Antibody (FABP2/7669), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP320667100UG
Description
FABP2/I-FABP Monoclonal antibody specifically detects FABP2/I-FABP in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
FABP2/I-FABP | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 μg/mL | |
FABPI, fatty acid binding protein 2, intestinal, Fatty acid-binding protein 2, fatty acid-binding protein, intestinal, I-FABPMGC133132, Intestinal-type fatty acid-binding protein | |
Recombinant full length human FABP2/I-FABP protein | |
100 μg | |
Cholesterol Metabolism, Lipid and Metabolism | |
2169 | |
Store at 4&drg;C. Do not freeze. | |
IgG |
Immunohistochemistry (Paraffin) | |
FABP2/7669 | |
Unconjugated | |
10mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction