Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FABP2/I-FABP Antibody (OTI2C4), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270699AF594
Description
FABP2/I-FABP Monoclonal specifically detects FABP2/I-FABP in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
FABP2/I-FABP | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
FABP2 | |
Full length recombinant protein of human FABP2/I-FABP (NP_000125) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG2b |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI2C4 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
FABPI, fatty acid binding protein 2, intestinal, Fatty acid-binding protein 2, fatty acid-binding protein, intestinal, I-FABPMGC133132, Intestinal-type fatty acid-binding protein | |
Mouse | |
Affinity Purified | |
Cholesterol Metabolism, Lipid and Metabolism | |
2169.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction