Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FABP2 Monoclonal Antibody (C1)
Mouse Monoclonal Antibody
$184.40 - $493.00
Specifications
| Antigen | FABP2 |
|---|---|
| Clone | C1 |
| Concentration | 1 mg/mL |
| Content And Storage | -20°C |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
PIMA541584
|
Invitrogen™
MA541584 |
200 μL |
Each for $493.00
|
|
|||||
|
PIMA543833
|
Invitrogen™
MA543833 |
20 μL |
Each for $184.40
|
|
|||||
Description
FABP2 Monoclonal Antibody for Western Blot, IHC (P)
The FABP2 gene encodes the intestinal fatty acid-binding protein 2 (I-FABP), which plays a significant role in lipid metabolism by facilitating the transport and intracellular trafficking of long-chain fatty acids in enterocytes, the absorptive cells in the intestine. FABP2 is involved in the uptake, metabolism, and transfer of fatty acids across cellular membranes, contributing to digestion and absorption processes in the intestine. This protein is crucial for efficient lipid metabolism, influencing fatty acid oxidation, and energy balance. Variations in the FABP2 gene have been associated with metabolic traits, such as insulin sensitivity, lipid profiles, and obesity, suggesting its impact on metabolic health. FABP2's function in intestinal lipid absorption and metabolism makes it a subject of interest in studies related to dietary effects on health and the development of metabolic disorders. Understanding FABP2's role is essential for insights into nutritional influences on health and possible therapeutic targets for metabolic diseases.Specifications
| FABP2 | |
| 1 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300 | |
| P02693 | |
| 25598 | |
| Recombinant protein Fatty Acid Binding Protein 2, Intestinal. The antigen corresponds to amino acid range 2-132 of the target protein. | |
| Antibody |
| C1 | |
| -20°C | |
| Monoclonal | |
| Liquid | |
| IgG2b κ | |
| Rat | |
| FABP2 | |
| FABP; Fabp2; Fabpi; fatty acid binding protein 1; fatty acid binding protein 2; fatty acid binding protein 2, intestinal; fatty acid-binding protein 2; Fatty acid-binding protein, intestinal; I-FABP; intestinal fatty acid binding protein; intestinal fatty acid binding protein 2; intestinal-type fatty acid-binding protein; MGC133132 | |
| FABP2 | |
| Primary | |
| Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title