Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ FABP2, Mouse, Unlabeled, Clone: 4F6.1,

Catalog No. MABS1694MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABS1694MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1694MI Supplier MilliporeSigma™ Supplier No. MABS1694

Mouse Monoclonal Antibody

Fatty acid-binding protein, intestinal (UniProt P12104; also known as Fatty acid-binding protein 2, I-FABP, Intestinal-type fatty acid-binding protein) is encoded by the FABP2 (also known as FABPI) gene (Gene ID 2169) in human. The intracellular lipid-binding protein (iLBP) multigene family includes 12 fatty acid-binding protein (FABPs), 4 retinol-binding proteins (RBPs) and 2 cellular retinoic acid-binding proteins (CRABPs). FABPs are 14-16kDa cytosolic proteins that bind fatty acids (FAs) and other hydrophobic ligands, such as eicosanoids and retinoids. FABPs are either numbered by the Hertzel and Bernlohr nomenclature to indicate their chronological order of discovery or named according to the tissue where each FABP was first isolated. In addition to transporting and sequestering long-chain FAs, eicosanoids, bile salts, and other hydrophobic ligands for storage and metabolism, FABPs are long recognized for their role in gene transcription regulation via lipid-mediated activation of nuclear hormone receptors (NHRs). FABP1 and FABP2, for example, are shown to bind a range of PPAR agonists, including long chain fatty acids (LCFA) and drugs. FABPs play differential and ligand-specific roles in NHR activation, and the patterns of drug interaction with intracellular binding proteins such as FABPs may govern tissue-specific actions of drugs that target NHRs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FABP2
Applications Western Blot
Classification Monoclonal
Clone 4F6.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse IgG1 in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene FABP2, FABPI
Gene Accession No. P12104
Host Species Mouse
Immunogen GST-tagged recombinant human FABP2.
Purification Method Protein G Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_000125
Test Specificity Clone 4F6.1 targets an epitope conserved among human, mouse, and rat species.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.