Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human FABP3 (cardiac) ELISA Kit

Catalog No. EHFABP3
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EHFABP3 96 Tests
EHFABP3X10 10 x 96 Tests
EHFABP3X5 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EHFABP3 Supplier Invitrogen™ Supplier No. EHFABP3
Add to Cart
Add to Cart

ELISA

The Human FABP3 (cardiac) ELISA quantitates Hu FABP3 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu FABP3. Principle of the method The Human FABP3 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Fatty acid binding proteins (FABP) are small (approximately 13-14 kDa) intracellular proteins with a high degree of tissue specificity. FABPs are a class of cytoplasmic proteins that bind long chain fatty acids. They are abundantly present in various cell types and seem to play an important role in the intracellular utilization of fatty acids. There are at least six distinct types of FABP, each showing a specific pattern of tissue expression. FABP leaks, due to its small size, rapidly out of is chemically damaged dying cells leading to a rise in serum levels. Liver FABP is a sensitive marker for cell damage of liver cells in vitro and in vivo. Is chemically damaged tissues are characterized histologically by absence (or low presence) of FABP facilitating recognition of such areas. Next to this L FABP is a marker for rapid hepatocyte lysis in vitro (as for example in toxicology assays) and for detection of liver damage during and after transplantation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P05413
Assay Range 1.5-200 ng/mL
Assay Sensitivity 1.5 ng/mL
Calibration Range 1.5 ng/mL to 200 ng/mL
Content And Storage Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8°C. See product manual for detailed contents and storage conditions for maximum stability.
Assay Solid-phase sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Other Proteins
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA pair antibody detects human FABP3. Other species not determined.
Sufficient For 96 assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 422 protein; Cardiac Fatty Acid Binding Protein; FABP 3; FABP11; Fabp3; Fabph1; Fabph-1; Fabph4; Fabph-4; fatty acid binding protein 11; Fatty acid binding protein 11 (FABP11); fatty acid binding protein 3; Fatty acid binding protein 3 heart; fatty acid binding protein 3, muscle and heart; Fatty acid binding protein heart; fatty acid binding protein heart 1; fatty acid binding protein heart 4; fatty acid-binding protein 3; Fatty acid-binding protein, heart; heart FABP; heart fatty acid binding protein; Heart type fatty acid binding protein; heart-type fatty acid-binding protein; H-FABP; M FABP; Mammary derived growth inhibitor; Mammary-derived growth inhibitor; MDGI; M-FABP; muscle and heart (mammary derived growth inhibitor); Muscle fatty acid binding protein; Muscle fatty acid-binding protein; Muscle O-FABP; Mylein protein P2 homolog / adopocyte; O-FABP422 protein; Cardiac Fatty Acid Binding Protein; FABP 3; FABP11; Fabp3; Fabph1; Fabph-1; Fabph4; Fabph-4; fatty acid binding protein 11; Fatty acid binding protein 11 (FABP11); fatty acid binding protein 3; Fatty acid binding protein 3 heart; fatty acid binding protein 3, muscle and heart; Fatty acid binding protein heart; fatty acid binding protein heart 1; fatty acid binding protein heart 4; fatty acid-binding protein 3; Fatty acid-binding protein, heart; heart FABP; heart fatty acid binding protein; Heart type fatty acid binding protein; heart-type fatty acid-binding protein; H-FABP; M FABP; Mammary derived growth inhibitor; Mammary-derived growth inhibitor; MDGI; M-FABP; muscle and heart (mammary derived growth inhibitor); Muscle fatty acid binding protein; Muscle fatty acid-binding protein; Muscle O-FABP; Mylein protein P2 homolog / adopocyte; O-FABP;
Gene ID (Entrez) 2170
Gene Symbol FABP3
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Research Discipline Other Research
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Synonym FABP11, H-FABP, M-FABP, MDGI, O-FABP
Target FABP3
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.