Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FABP3/H-FABP Antibody (9F10), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP260234JF646
This item is not returnable.
View return policy
Description
FABP3/H-FABP Monoclonal antibody specifically detects FABP3/H-FABP in Human samples. It is validated for Western Blot, ELISASpecifications
FABP3/H-FABP | |
Monoclonal | |
Janelia Fluor 646 | |
FABP11, fatty acid binding protein 11, fatty acid binding protein 3, muscle and heart (mammary-derived growthinhibitor), Fatty acid-binding protein 3, Fatty acid-binding protein 3, muscle, fatty acid-binding protein, heart, Heart-type fatty acid-binding protein, H-FABPM-FABP, Mammary-derived growth inhibitor, MDGI, Muscle fatty acid-binding protein, O-FABP | |
Recombinant human FABP3/H-FABP (1-133aa) purified from E. coli | |
0.1 mL | |
Cell Cycle and Replication, Cholesterol Metabolism, Lipid and Metabolism | |
2170 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, ELISA | |
9F10 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction