Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FABP4/A-FABP Antibody (1H4), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | FABP4/A-FABP |
---|---|
Clone | 1H4 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Description
FABP4/A-FABP Monoclonal antibody specifically detects FABP4/A-FABP in Human samples. It is validated for ELISASpecifications
FABP4/A-FABP | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Adipocyte lipid-binding protein, Adipocyte-type fatty acid-binding protein, A-FABPAFABP, ALBP, aP2, fatty acid binding protein 4, adipocyte, Fatty acid-binding protein 4, fatty acid-binding protein, adipocyte | |
A synthesized peptide derived from Human FABP4/A-FABP [UniProt P15090] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
1H4 | |
ELISA | |
Unconjugated | |
Rabbit | |
Cancer, Phospho Specific, Stem Cell Markers | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
2167 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title