Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FABP4/A-FABP Rabbit anti-Human, Clone: 1, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $548.00
Specifications
Antigen | FABP4/A-FABP |
---|---|
Clone | 1 |
Dilution | Flow Cytometry 1:25-1:100, Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000-1:4000 |
Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
FABP4/A-FABP Monoclonal specifically detects FABP4/A-FABP in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
FABP4/A-FABP | |
Flow Cytometry 1:25-1:100, Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000-1:4000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Adipocyte lipid-binding protein, Adipocyte-type fatty acid-binding protein, A-FABPAFABP, ALBP, aP2, fatty acid binding protein 4, adipocyte, Fatty acid-binding protein 4, fatty acid-binding protein, adipocyte | |
Recombinant Human FABP4/A-FABP protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
1 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cancer, Phospho Specific, Stem Cell Markers | |
0.2 um filtered solution in PBS | |
2167 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title