Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FABP4 Recombinant Rabbit Monoclonal Antibody (HL1521)
GREENER_CHOICE

Catalog No. PIMA547077
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547077 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547077 Supplier Invitrogen™ Supplier No. MA547077
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

FABP4 belongs to the fatty acid binding protein (FABP) family. FABPs contribute to the transport of fatty acids and other lipids in various cellular pathways. FABP4 is expressed in adipocytes, macrophages, monocyte-derived dendritic cells, and myoepithelial cells. It delivers long-chain fatty acids and retinoic acid to their cognate receptors in the nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FABP4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL1521
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene FABP4
Gene Accession No. P04117, P15090, P70623
Gene Alias 3T3-L1 lipid-binding protein; 422/aP2; adipocyte fatty acid binding protein; Adipocyte lipid binding protein; adipocyte lipid-binding protein; adipocyte protein aP2; adipocyte-type fatty acid-binding protein; AFABP; A-FABP; ALBP; ALBP/Ap2; Ap2; epididymis secretory protein Li 104; FABP4; fatty acid binding protein 4; Fatty acid binding protein 4 adipocyte; fatty acid binding protein 4, adipocyte; fatty acid-binding protein 4; Fatty acid-binding protein, adipocyte; HEL S 104; HEL-S-104; Lbpl; myelin P2 protein homolog; P15; P2 adipocyte protein; Protein 422
Gene Symbols FABP4
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human FABP4. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11770, 2167, 79451
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.