Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FABP4 Mouse anti-Human, Clone: 5C1, Bioss
SDP

Catalog No. 501979825
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-197-9825 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-197-9825 Supplier Bioss Supplier No. BSM51247M
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FABP4 belongs to the fatty acid binding protein (FABP) family. FABPs contribute to the transport of fatty acids and other lipids in various cellular pathways. FABP4 is expressed in adipocytes, macrophages, monocyte-derived dendritic cells, and myoepithelial cells. It delivers long-chain fatty acids and retinoic acid to their cognate receptors in the nucleus.

Specifications

Antigen FABP4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 5C1
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 40% glycerol and 0.05% sodium azide; pH
Gene FABP4
Gene Accession No. P15090
Gene Alias 3T3-L1 lipid-binding protein; 422/aP2; adipocyte fatty acid binding protein; Adipocyte lipid-binding protein; adipocyte protein aP2; Adipocyte-type fatty acid-binding protein; AFABP; A-FABP; ALBP; ALBP/Ap2; Ap2; epididymis secretory protein Li 104; Fabp4; fatty acid binding protein 4; fatty acid binding protein 4, adipocyte; Fatty acid-binding protein 4; Fatty acid-binding protein, adipocyte; HEL-S-104; Lbpl; Myelin P2 protein homolog; P15; P2 adipocyte protein; Protein 422
Gene Symbols FABP4
Host Species Mouse
Immunogen Purified His-tagged FABP4 protein was used to produced this monoclonal antibody.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2167
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.