Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FABP5/E-FABP Antibody (311215), DyLight 405, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB3077E
Description
FABP5/E-FABP Monoclonal antibody specifically detects FABP5/E-FABP in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF, Gene Knock-OutSpecifications
FABP5/E-FABP | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF, Gene Knock-Out | |
311215 | |
Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready, Knockout Validated | |
EFABP, E-FABPPAFABP, fatty acid binding protein 5 (psoriasis-associated), Fatty acid-binding protein 5, PA-FABPepidermal, Psoriasis-associated fatty acid-binding protein homolog | |
E. coli-derived recombinant human FABP5/E-FABP, Ala2-Glu135, Accession # Q01469 | |
0.1 mL | |
Stem Cell Markers | |
2171 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction