Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FABP5/E-FABP Rabbit anti-Human, Clone: FABP5/6353R, Novus Biologicals™

Rabbit Monoclonal Antibody
$274.50 - $527.00
Specifications
Antigen | FABP5/E-FABP |
---|---|
Clone | FABP5/6353R |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 1-2 ug/ml, Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml |
Applications | Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) |
Description
FABP5/E-FABP Monoclonal antibody specifically detects FABP5/E-FABP in Human samples. It is validated for Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
FABP5/E-FABP | |
0.2 mg/ml | |
Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Stem Cell Markers | |
10 mM PBS with 0.05% BSA | |
EFABP, E-FABPPAFABP, fatty acid binding protein 5 (psoriasis-associated), Fatty acid-binding protein 5, PA-FABPepidermal, Psoriasis-associated fatty acid-binding protein homolog | |
Recombinant human full-length FABP5/E-FABP protein (Uniprot: Q01469) | |
Primary | |
Store at 4°C. |
FABP5/6353R | |
Western Blot 1-2 ug/ml, Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Q01469 | |
2171 | |
IgG | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title