Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FABP5 Monoclonal Antibody (311215)
GREENER_CHOICE

Catalog No. PIMA524029
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524029 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524029 Supplier Invitrogen™ Supplier No. MA524029
Add to Cart
Add to Cart

Rat Monoclonal Antibody

In direct ELISAs and Western blots, no cross-reactivity with recombinant human FABP1, 2, 3, 4, 6, 7, or 9 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

FABP5 (fatty acid binding protein 5 (psoriasis-associated)) is a proteinthat in humans is encoded by theFABP5 gene, also known as PAFABP, EFABP, E-FABP, KFABP, PA-FABP, Epidermal-type fatty acid-binding protein, Fatty acid-binding protein 5, Psoriasis-associated fatty acid-binding protein homolog. It is a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. The PAFABP cDNA encodes a 135-amino acid protein with molecular weight 15,164. This gene encodes the fatty acid binding protein found in epidermal cells, and was first identified as being upregulated inpsoriasistissue. It is though that FABPs roles include fatty acid uptake, transport, and metabolism. Kaczocha et al. (2009) identifiedFABP5and FABP7 as cytosolic proteins that transport AEA from the plasma membrane to subcellular fatty acid amide hydrolase, where it is hydrolyzed and inactivated. FABP3 did not show this specific transport function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FABP5
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 311215
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative; pH 7.4
Gene Fabp5
Gene Accession No. Q01469
Gene Alias C-FABP; Cutaneous fatty acid-binding protein; DA11; EFABP; E-FABP; E-FABPPAFABP; Epidermal-type fatty acid-binding protein; epithelial fatty acid-binding protein; Fabp5; Fabpe; fatty acid binding protein 5; fatty acid binding protein 5 (psoriasis-associated); fatty acid binding protein 5, epidermal; fatty acid-binding protein 5; Fatty acid-binding protein, epidermal; fatty acid-binding protein, epidermal; uncharacterized protein LOC16592; keratinocyte lipid-binding protein; KFABP; Klbp; Mal1; PAFABP; PA-FABP; PA-FABPepidermal; Psoriasis-associated fatty acid-binding protein homolog
Gene Symbols Fabp5
Host Species Rat
Immunogen E. coli-derived recombinant human FABP5/E-FABP Ala2-Glu135.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2171
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.