Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FABP5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120059
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120059 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120059 Supplier Invitrogen™ Supplier No. PA5120059
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: 293T. The target is usually found in the following locations: Cytoplasm. Immunogen sequence: MATVQQLEGR WRLVDSKGFD EYMKELGVGI ALRKMGAMAK PDCIITCDGK NLTIKTESTL KTTQFSCTLG EKFEETTADG RKTQTVCNFT DGALVQHQEW.

FABP5 (fatty acid binding protein 5 (psoriasis-associated)) is a proteinthat in humans is encoded by theFABP5 gene, also known as PAFABP, EFABP, E-FABP, KFABP, PA-FABP, Epidermal-type fatty acid-binding protein, Fatty acid-binding protein 5, Psoriasis-associated fatty acid-binding protein homolog. It is a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. The PAFABP cDNA encodes a 135-amino acid protein with molecular weight 15,164. This gene encodes the fatty acid binding protein found in epidermal cells, and was first identified as being upregulated inpsoriasistissue. It is though that FABPs roles include fatty acid uptake, transport, and metabolism. Kaczocha et al. (2009) identifiedFABP5and FABP7 as cytosolic proteins that transport AEA from the plasma membrane to subcellular fatty acid amide hydrolase, where it is hydrolyzed and inactivated. FABP3 did not show this specific transport function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FABP5
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Fabp5
Gene Accession No. Q01469
Gene Alias C-FABP; Cutaneous fatty acid-binding protein; DA11; EFABP; E-FABP; E-FABPPAFABP; Epidermal-type fatty acid-binding protein; epithelial fatty acid-binding protein; Fabp5; Fabpe; fatty acid binding protein 5; fatty acid binding protein 5 (psoriasis-associated); fatty acid binding protein 5, epidermal; fatty acid-binding protein 5; Fatty acid-binding protein, epidermal; fatty acid-binding protein, epidermal; uncharacterized protein LOC16592; keratinocyte lipid-binding protein; KFABP; Klbp; Mal1; PAFABP; PA-FABP; PA-FABPepidermal; Psoriasis-associated fatty acid-binding protein homolog
Gene Symbols Fabp5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human FABP5 (NP_001435.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2171
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.