Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FABP6 Antibody (009), Alexa Fluor™ 700, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290126AF700
Description
FABP6 Monoclonal specifically detects FABP6 in Human samples. It is validated for ELISA.Specifications
| FABP6 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 9 | |
| ELISA | |
| fatty acid binding protein 6, ileal, Fatty acid-binding protein 6, gastrotropin, I-15PIntestinal 15 kDa protein, I-BABPIntestinal bile acid-binding protein, I-BALB, I-BAP, ILBP3, ILBPGT, ileal bile acid binding protein, ILLBPIleal lipid-binding protein, illeal lipid-binding protein | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human FABP6 (AAH22489.1; Met1-Ala128). | |
| 0.1 mL | |
| Biologically Active Proteins, Cell Cycle and Replication | |
| 2172 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction